Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_004322.3 | 624 | Silent Mutation | CGC,CGT | R96R | NP_004313.1 |
NM_032989.2 | 624 | Silent Mutation | CGC,CGT | R96R | NP_116784.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001170726.1 | 624 | Intron | NP_001164197.1 | ||
NM_001170880.1 | 624 | Intron | NP_001164351.1 | ||
NM_001170881.1 | 624 | Intron | NP_001164352.1 | ||
NM_001177358.1 | 624 | Intron | NP_001170829.1 | ||
NM_020155.3 | 624 | Intron | NP_064540.3 | ||
XM_005274100.2 | 624 | Intron | XP_005274157.1 | ||
XM_005274101.2 | 624 | Intron | XP_005274158.1 | ||
XM_005274102.2 | 624 | Intron | XP_005274159.1 | ||
XM_005274104.2 | 624 | Intron | XP_005274161.1 | ||
XM_011545168.2 | 624 | Intron | XP_011543470.1 | ||
XM_011545169.1 | 624 | Intron | XP_011543471.1 | ||
XM_011545170.2 | 624 | Intron | XP_011543472.1 | ||
XM_011545171.2 | 624 | Intron | XP_011543473.1 | ||
XM_011545172.2 | 624 | Intron | XP_011543474.1 | ||
XM_017018014.1 | 624 | Intron | XP_016873503.1 | ||
XM_017018015.1 | 624 | Intron | XP_016873504.1 | ||
XM_017018016.1 | 624 | Intron | XP_016873505.1 |