Product Details
- SNP ID
-
rs2026316
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:10441705 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGTTACTTGCATTATCCTCTTTCCA[G/T]TGTTGAATCTGACGTGTTTGTTTTT
- Phenotype
-
MIM: 604896
MIM: 615958
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MKKS
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs188349794] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MKKS
- Gene Name
- McKusick-Kaufman syndrome
There are no transcripts associated with this gene.
- Gene
- SLX4IP
- Gene Name
- SLX4 interacting protein
View Full Product Details