Product Details

SNP ID
rs12599876
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:68359502 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCGCCAGGCTCCCTGGGCTCTGCAG[C/T]GTTGTCTGCCCCAGCACAGGGCCAG
Phenotype
MIM: 610087 MIM: 605777
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRMT7 PubMed Links

Gene Details

Gene
PRMT7
Gene Name
protein arginine methyltransferase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184824.1 4494 Intron NP_001171753.1
NM_001290018.1 4494 Intron NP_001276947.1
NM_019023.2 4494 Intron NP_061896.1
XM_011523112.2 4494 Intron XP_011521414.1
XM_011523113.2 4494 Intron XP_011521415.1
XM_011523115.2 4494 Intron XP_011521417.1
XM_011523116.2 4494 Intron XP_011521418.1
XM_011523121.2 4494 Intron XP_011521423.1
XM_011523124.2 4494 Intron XP_011521426.1
XM_011523125.2 4494 Intron XP_011521427.1
XM_011523126.2 4494 Intron XP_011521428.1
XM_011523128.2 4494 Intron XP_011521430.1
XM_011523131.2 4494 Intron XP_011521433.1
XM_017023290.1 4494 Intron XP_016878779.1
XM_017023291.1 4494 Intron XP_016878780.1
XM_017023292.1 4494 Intron XP_016878781.1
XM_017023293.1 4494 Intron XP_016878782.1
XM_017023294.1 4494 Intron XP_016878783.1
XM_017023295.1 4494 Intron XP_016878784.1
XM_017023296.1 4494 Intron XP_016878785.1
XM_017023297.1 4494 Intron XP_016878786.1
XM_017023298.1 4494 Intron XP_016878787.1
XM_017023299.1 4494 Intron XP_016878788.1
XM_017023300.1 4494 Intron XP_016878789.1
XM_017023301.1 4494 Intron XP_016878790.1
XM_017023302.1 4494 Intron XP_016878791.1
XM_017023303.1 4494 Intron XP_016878792.1
XM_017023304.1 4494 Intron XP_016878793.1
XM_017023305.1 4494 Intron XP_016878794.1
XM_017023306.1 4494 Intron XP_016878795.1
XM_017023307.1 4494 Intron XP_016878796.1
XM_017023308.1 4494 Intron XP_016878797.1
XM_017023309.1 4494 Intron XP_016878798.1
XM_017023310.1 4494 Intron XP_016878799.1
XM_017023311.1 4494 Intron XP_016878800.1
XM_017023312.1 4494 Intron XP_016878801.1
XM_017023313.1 4494 Intron XP_016878802.1
XM_017023314.1 4494 Intron XP_016878803.1
XM_017023315.1 4494 Intron XP_016878804.1
XM_017023316.1 4494 Intron XP_016878805.1
Gene
SMPD3
Gene Name
sphingomyelin phosphodiesterase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018667.3 4494 UTR 3 NP_061137.1
XM_005256031.3 4494 UTR 3 XP_005256088.1
XM_005256032.3 4494 UTR 3 XP_005256089.1
XM_011523207.1 4494 UTR 3 XP_011521509.1
XM_011523208.2 4494 UTR 3 XP_011521510.1
XM_011523209.2 4494 UTR 3 XP_011521511.1
XM_011523210.2 4494 Intron XP_011521512.1
XM_017023405.1 4494 UTR 3 XP_016878894.1
XM_017023406.1 4494 UTR 3 XP_016878895.1
XM_017023407.1 4494 UTR 3 XP_016878896.1
XM_017023408.1 4494 UTR 3 XP_016878897.1

View Full Product Details