Product Details

SNP ID
rs7248579
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1910361 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCTGGAAGCCAGGCGCCGCCTG[C/T]CCCCGCTTCTGCTTCCAGGCTCTGG
Phenotype
MIM: 615302 MIM: 613764
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ADAT3 PubMed Links

Gene Details

Gene
ADAT3
Gene Name
adenosine deaminase, tRNA specific 3
There are no transcripts associated with this gene.

Gene
SCAMP4
Gene Name
secretory carrier membrane protein 4
There are no transcripts associated with this gene.

View Full Product Details