Product Details

SNP ID
hCV11698022
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.13:51935599 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
TCCACCTGAGGGGACTCACCACTTG[A/G]GCTGCAGGGATGAGAGCACCACAGA
Phenotype
MIM: 606882
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATP7B PubMed Links

Gene Details

Gene
ATP7B
Gene Name
ATPase copper transporting beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000053.3 3654 Missense Mutation CCC,CTC P1373L NP_000044.2
NM_001005918.2 3654 Missense Mutation CCC,CTC P1166L NP_001005918.1
NM_001243182.1 3654 Missense Mutation CCC,CTC P1262L NP_001230111.1

View Full Product Details