Product Details

SNP ID
rs1045692
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:120764465 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTGTGGATTTAATAAGTCACTT[C/T]ACACCACAGAAAATATTTAATAAAT
Phenotype
MIM: 608240
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
SPPL3 PubMed Links

Gene Details

Gene
SPPL3
Gene Name
signal peptide peptidase like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139015.4 2174 UTR 3 NP_620584.2
XM_011537925.2 2174 UTR 3 XP_011536227.1

View Full Product Details