Product Details
- SNP ID
-
rs3212797
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:17826977 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCCAAAGGACACAACTCCCATTCAG[T/C]GTATTTGTTTTTGTTTTTTTGAGAC
- Phenotype
-
MIM: 146738
MIM: 600173
- Polymorphism
- T/C, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
INSL3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3212798] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- INSL3
- Gene Name
- insulin like 3
There are no transcripts associated with this gene.
- Gene
- JAK3
- Gene Name
- Janus kinase 3
View Full Product Details