Product Details

SNP ID
rs1920220
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:123888158 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
AATTCTGATGTATGTTGTTCTTTCA[A/T]GTTTCATGTCACTCAGTGTGTATTA
Phenotype
MIM: 600922
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
CCDC14 PubMed Links

Gene Details

Gene
CCDC14
Gene Name
coiled-coil domain containing 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308317.1 Intron NP_001295246.1
NM_022757.4 Intron NP_073594.4
XM_005247710.4 Intron XP_005247767.1
XM_005247711.4 Intron XP_005247768.1
XM_005247713.2 Intron XP_005247770.1
XM_005247714.2 Intron XP_005247771.1
XM_005247715.4 Intron XP_005247772.1
XM_005247716.4 Intron XP_005247773.1
XM_006713731.3 Intron XP_006713794.1
XM_006713733.3 Intron XP_006713796.1
XM_011513081.2 Intron XP_011511383.1
XM_017007056.1 Intron XP_016862545.1
XM_017007057.1 Intron XP_016862546.1
XM_017007058.1 Intron XP_016862547.1
XM_017007059.1 Intron XP_016862548.1
XM_017007060.1 Intron XP_016862549.1
XM_017007061.1 Intron XP_016862550.1
Gene
MYLK
Gene Name
myosin light chain kinase
There are no transcripts associated with this gene.

View Full Product Details