Product Details

SNP ID
rs1804674
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:19454759 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAAATACACTCACCTCTTCAACC[T/C]TTTTGACAAGGGGACGTGAATTTCT
Phenotype
MIM: 601754
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
C22orf39 PubMed Links

Gene Details

Gene
C22orf39
Gene Name
chromosome 22 open reading frame 39
There are no transcripts associated with this gene.

Gene
UFD1L
Gene Name
ubiquitin fusion degradation 1 like (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001035247.2 1095 Intron NP_001030324.2
NM_005659.6 1095 Intron NP_005650.2
XM_011530361.2 1095 Missense Mutation AAG,AGG K275R XP_011528663.1

View Full Product Details