Product Details

SNP ID
rs2032934
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:157270139 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGAGGGAAGTAAATAGGACCTTCT[G/T]GCGAGATGCCTGAGAAAGACCCAGT
Phenotype
MIM: 606323
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CYFIP2 PubMed Links

Gene Details

Gene
CYFIP2
Gene Name
cytoplasmic FMR1 interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037333.2 Intron NP_001032410.1
NM_001291721.1 Intron NP_001278650.1
NM_001291722.1 Intron NP_001278651.1
NM_014376.3 Intron NP_055191.2
XM_011534516.2 Intron XP_011532818.1
XM_017009341.1 Intron XP_016864830.1
XM_017009342.1 Intron XP_016864831.1

View Full Product Details