Product Details

SNP ID
rs2023822
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:132698654 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTAAAAATAAATTCTATTACTCAG[A/G]AGCCTAAGAAGTTGGAAGACCTTAA
Phenotype
MIM: 604683
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIF3A PubMed Links

Gene Details

Gene
KIF3A
Gene Name
kinesin family member 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300791.1 Intron NP_001287720.1
NM_001300792.1 Intron NP_001287721.1
NM_007054.6 Intron NP_008985.3
XM_006714526.3 Intron XP_006714589.1
XM_017008995.1 Intron XP_016864484.1
XM_017008996.1 Intron XP_016864485.1

View Full Product Details