Product Details

SNP ID
rs12679128
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:52543171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAAATGCAATTTATACAAACAAT[C/T]GGGAGCATGTGCAACTCTTTCAAAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM150A PubMed Links

Gene Details

Gene
FAM150A
Gene Name
family with sequence similarity 150 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207413.3 Intron NP_997296.1

View Full Product Details