Product Details
- SNP ID
-
rs34872080
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:153338169 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATAGAAGAGTTGTGCAACATGGACC[A/C]TAAATGATCCGATTGTCTCTAGGAT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF275
PubMed Links
Gene Details
- Gene
- ZNF275
- Gene Name
- zinc finger protein 275
View Full Product Details