Product Details

SNP ID
rs12606958
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:75412190 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGAAGCCCAAATTCTCCAAGAGA[C/G]GGCAAGTACATGTGAAAACAAACAA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SMIM21 PubMed Links

Gene Details

Gene
SMIM21
Gene Name
small integral membrane protein 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037331.3 Intron NP_001032408.1
NM_001303482.1 Intron NP_001290411.1
XM_011525928.1 Intron XP_011524230.1

View Full Product Details