Product Details

SNP ID
rs1865723
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:75417525 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGCTAAGGAAGAGAGAATATAGCT[A/G]ATGTAGAATAAACTCCCTATAGTAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SMIM21 PubMed Links

Gene Details

Gene
SMIM21
Gene Name
small integral membrane protein 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037331.3 1068 Intron NP_001032408.1
NM_001303482.1 1068 UTR 3 NP_001290411.1
XM_011525928.1 1068 UTR 3 XP_011524230.1

View Full Product Details