Product Details

SNP ID
rs4838754
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:133075211 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCTCTGCAGCCCACAGCACCCCC[T/C]GCCGAGGCCCAGCAGCTGACCTCTG
Phenotype
MIM: 612302
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
ADGRA1 PubMed Links

Gene Details

Gene
ADGRA1
Gene Name
adhesion G protein-coupled receptor A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083909.2 Intron NP_001077378.1
NM_001291085.1 Intron NP_001278014.1
XM_005252695.1 Intron XP_005252752.1
XM_011540273.1 Intron XP_011538575.1
XM_017016779.1 Intron XP_016872268.1

View Full Product Details