Product Details
- SNP ID
-
rs10208406
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:10057488 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGTTCCTGAGATGCTCCTGCTCTGC[C/T]GTGTTCTAAGGACCTGCGAGCTGGA
- Phenotype
-
MIM: 603301
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CYS1
PubMed Links
Gene Details
- Gene
- CYS1
- Gene Name
- cystin 1
- Gene
- KLF11
- Gene Name
- Kruppel like factor 11
There are no transcripts associated with this gene.
View Full Product Details