Product Details

SNP ID
rs17690965
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:132694975 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCACCTCAAGAGTACTCCACCTTCA[C/G]TCACCCCTACTCTATACACAGCTTT
Phenotype
MIM: 604683
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KIF3A PubMed Links

Gene Details

Gene
KIF3A
Gene Name
kinesin family member 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300791.1 3999 UTR 3 NP_001287720.1
NM_001300792.1 3999 UTR 3 NP_001287721.1
NM_007054.6 3999 UTR 3 NP_008985.3
XM_006714526.3 3999 Intron XP_006714589.1
XM_017008995.1 3999 Intron XP_016864484.1
XM_017008996.1 3999 Intron XP_016864485.1

View Full Product Details