Product Details

SNP ID
rs13143315
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:159117997 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCAAGGATTGATGAAATTTTCACC[A/G]TGGGTGGGTATTGTAATATTACCAA
Phenotype
MIM: 609530
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAPGEF2 PubMed Links

Gene Details

Gene
RAPGEF2
Gene Name
Rap guanine nucleotide exchange factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014247.2 Intron NP_055062.1
XM_005263358.3 Intron XP_005263415.1
XM_005263359.3 Intron XP_005263416.1
XM_005263360.3 Intron XP_005263417.1
XM_005263361.3 Intron XP_005263418.1
XM_006714420.3 Intron XP_006714483.1
XM_006714421.3 Intron XP_006714484.1
XM_006714422.3 Intron XP_006714485.1
XM_011532425.2 Intron XP_011530727.1
XM_011532426.2 Intron XP_011530728.1
XM_011532427.2 Intron XP_011530729.1
XM_017008856.1 Intron XP_016864345.1
XM_017008857.1 Intron XP_016864346.1
XM_017008858.1 Intron XP_016864347.1
XM_017008859.1 Intron XP_016864348.1

View Full Product Details