Product Details

SNP ID
rs174923
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:29814709 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTTGTGAAGAGTTGTTTTGGGGGA[C/T]GAGAGATAGTTATGTAAGTGACAAT
Phenotype
MIM: 612432
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WIPF3 PubMed Links

Gene Details

Gene
WIPF3
Gene Name
WAS/WASL interacting protein family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080529.2 Intron NP_001073998.2
XM_017012522.1 Intron XP_016868011.1

View Full Product Details