Product Details

SNP ID
rs2041556
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:15118063 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTTATTGCAAACATAATTTCCCT[G/T]TGTAATGTTGCTTGAAAAGTTGACC
Phenotype
MIM: 613738
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AGMO PubMed Links

Gene Details

Gene
AGMO
Gene Name
alkylglycerol monooxygenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004320.1 Intron NP_001004320.1
XM_006715730.1 Intron XP_006715793.1
XM_006715731.3 Intron XP_006715794.1
XM_011515402.2 Intron XP_011513704.1
XM_017012204.1 Intron XP_016867693.1

View Full Product Details