Product Details

SNP ID
rs7087038
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:87106372 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCCACAGAAAATAACTGACTTGCT[C/G]TCTGCTGTCTTACTGTGATAATATA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM35A PubMed Links

Gene Details

Gene
FAM35A
Gene Name
family with sequence similarity 35 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019054.2 Intron NP_061927.2
XM_005269920.3 Intron XP_005269977.1
XM_006717903.3 Intron XP_006717966.1
XM_006717905.3 Intron XP_006717968.1
XM_011539876.2 Intron XP_011538178.1
XM_011539877.2 Intron XP_011538179.1
XM_011539878.2 Intron XP_011538180.1
XM_011539879.2 Intron XP_011538181.1
XM_011539881.2 Intron XP_011538183.1
XM_011539882.2 Intron XP_011538184.1
XM_011539883.2 Intron XP_011538185.1
XM_017016349.1 Intron XP_016871838.1
XM_017016350.1 Intron XP_016871839.1
XM_017016351.1 Intron XP_016871840.1
XM_017016352.1 Intron XP_016871841.1
XM_017016353.1 Intron XP_016871842.1

View Full Product Details