Product Details

SNP ID
rs1007127
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:70049034 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GCAGAAATGTCTCAGGTGAACAGGA[A/G]GTAAAAAGAGAAATTGAAAGCACTT
Phenotype
MIM: 612879
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAMDC2 PubMed Links

Gene Details

Gene
MAMDC2
Gene Name
MAM domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153267.4 Intron NP_694999.3
XM_011518471.1 Intron XP_011516773.1
XM_011518475.2 Intron XP_011516777.1
XM_017014587.1 Intron XP_016870076.1

View Full Product Details