Product Details

SNP ID
rs11586958
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:41508493 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAGCAGCCAGGAGCTGCTGGCTAA[C/G]CCTGATTCCCCAGAACCCTGGAGCA
Phenotype
MIM: 606649
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HIVEP3 PubMed Links

Gene Details

Gene
HIVEP3
Gene Name
human immunodeficiency virus type I enhancer binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127714.2 10191 UTR 3 NP_001121186.1
NM_024503.4 10191 UTR 3 NP_078779.2
XM_011541884.2 10191 UTR 3 XP_011540186.1
XM_011541885.2 10191 Intron XP_011540187.1
XM_011541886.2 10191 Intron XP_011540188.1
XM_011541887.2 10191 Intron XP_011540189.1
XM_017001992.1 10191 UTR 3 XP_016857481.1
XM_017001993.1 10191 UTR 3 XP_016857482.1
XM_017001994.1 10191 UTR 3 XP_016857483.1

View Full Product Details