Product Details

SNP ID
rs12012897
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:80363183 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGCTTTTCTGTGTCATTGGCAAGC[C/T]TAGATAGGGTCTTTGTTAACTGAAT
Phenotype
MIM: 300976
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM46D PubMed Links

Gene Details

Gene
FAM46D
Gene Name
family with sequence similarity 46 member D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170574.1 Intron NP_001164045.1
NM_152630.4 Intron NP_689843.1

View Full Product Details