Product Details
- SNP ID
-
rs2240123
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:48075934 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- ACAGCTGCTAATATCAGGGAAGAAG[C/T]AGAACAAAACTACTTTGACAGTAGT
- Phenotype
-
MIM: 604511
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CBX1
PubMed Links
Gene Details
- Gene
- CBX1
- Gene Name
- chromobox 1
View Full Product Details