Product Details

SNP ID
rs1909979
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:75481796 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CACTGTACTCACCCCAGTTACATTT[C/T]AGATGAACCCCCTATTGTTTAATGT
Phenotype
MIM: 602692
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLIPR1 PubMed Links

Gene Details

Gene
GLIPR1
Gene Name
GLI pathogenesis related 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006851.2 Intron NP_006842.2
XM_011537775.2 Intron XP_011536077.1
XM_017018721.1 Intron XP_016874210.1

View Full Product Details