Product Details
- SNP ID
-
rs2855
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:16258078 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAAATGTTGGGTACTTTTTCCAAG[A/T]AAAATGTTTCTGAATGTGCACACTA
- Phenotype
-
MIM: 608959
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
DPH3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116028960] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DPH3
- Gene Name
- diphthamide biosynthesis 3
- Gene
- OXNAD1
- Gene Name
- oxidoreductase NAD binding domain containing 1
There are no transcripts associated with this gene.
View Full Product Details