Product Details
- SNP ID
-
rs1971476
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:113494637 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATTGGGATGATAACAAAAAGTTCTA[A/G]AGATCTAGGTATTGTGGCATATATA
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DCUN1D2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116043056] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DCUN1D2
- Gene Name
- defective in cullin neddylation 1 domain containing 2
There are no transcripts associated with this gene.
- Gene
- TMCO3
- Gene Name
- transmembrane and coiled-coil domains 3
View Full Product Details