Product Details

SNP ID
rs79657230
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44374673 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCACACTCACCTGAGCTTCCCCTC[A/G]GGGCAGGCTGAGCGGGGGCACCGCA
Phenotype
MIM: 607759
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITGA2B PubMed Links

Gene Details

Gene
ITGA2B
Gene Name
integrin subunit alpha 2b
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000419.4 3057 Nonsense Mutation CGA,TGA R977* NP_000410.2
XM_011524749.1 3057 Intron XP_011523051.1
XM_011524750.1 3057 Nonsense Mutation CGA,TGA R977* XP_011523052.1

View Full Product Details