Product Details
- SNP ID
-
rs2048228
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:99112451 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATATGTTTGTTTGAGAAGATACTGT[C/T]GACTTCAAATAGTACATTTTGAGGA
- Phenotype
-
MIM: 606087
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SYNM
PubMed Links
Gene Details
- Gene
- SYNM
- Gene Name
- synemin
View Full Product Details