Product Details

SNP ID
rs1811729
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:80780154 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTCGCCTCCCAGTGCCTCCGGGA[C/G]AGCGCTAGGCACCCAGCGACGCGCT
Phenotype
MIM: 608366
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CEMIP PubMed Links

Gene Details

Gene
CEMIP
Gene Name
cell migration inducing hyaluronan binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001293298.1 Intron NP_001280227.1
NM_001293304.1 Intron NP_001280233.1
NM_018689.2 Intron NP_061159.1

View Full Product Details