Product Details

SNP ID
rs13014
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:17201424 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGGAGCTATCTGTATATGACACAC[A/G]CATTATAAACAAATAACCTGGAACA
Phenotype
MIM: 607273 MIM: 612935 MIM: 614960
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FLCN PubMed Links

Gene Details

Gene
FLCN
Gene Name
folliculin
There are no transcripts associated with this gene.

Gene
MPRIP
Gene Name
myosin phosphatase Rho interacting protein
There are no transcripts associated with this gene.

Gene
PLD6
Gene Name
phospholipase D family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178836.3 2148 UTR 3 NP_849158.2
XM_017024310.1 2148 UTR 3 XP_016879799.1

View Full Product Details