Product Details

SNP ID
hCV11958118
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:46235395 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTAGACACGTTCCTGTCGCCTCT[G/A]ACAGCTGCAGTTGCTCCCTCATCAT
Phenotype
MIM: 600909 MIM: 603294
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
LSS PubMed Links

Gene Details

Gene
LSS
Gene Name
lanosterol synthase (2,3-oxidosqualene-lanosterol cyclase)
There are no transcripts associated with this gene.

Gene
MCM3AP
Gene Name
minichromosome maintenance complex component 3 associated protein
There are no transcripts associated with this gene.

Gene
MCM3AP-AS1
Gene Name
MCM3AP antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details