Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001271844.1 | 7455 | UTR 3 | NP_001258773.1 | ||
NM_001271845.1 | 7455 | UTR 3 | NP_001258774.1 | ||
NM_001271846.1 | 7455 | Intron | NP_001258775.1 | ||
NM_019020.3 | 7455 | UTR 3 | NP_061893.2 | ||
XM_005257049.2 | 7455 | UTR 3 | XP_005257106.1 | ||
XM_005257050.3 | 7455 | UTR 3 | XP_005257107.1 | ||
XM_005257052.3 | 7455 | Intron | XP_005257109.1 | ||
XM_006721694.3 | 7455 | UTR 3 | XP_006721757.1 | ||
XM_006721695.3 | 7455 | Intron | XP_006721758.1 | ||
XM_011524322.2 | 7455 | UTR 3 | XP_011522624.1 | ||
XM_011524323.2 | 7455 | UTR 3 | XP_011522625.2 | ||
XM_011524324.2 | 7455 | UTR 3 | XP_011522626.1 | ||
XM_017024185.1 | 7455 | Intron | XP_016879674.1 | ||
XM_017024186.1 | 7455 | UTR 3 | XP_016879675.1 | ||
XM_017024187.1 | 7455 | UTR 3 | XP_016879676.1 | ||
XM_017024188.1 | 7455 | UTR 3 | XP_016879677.1 |