Product Details

SNP ID
rs11213
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:79932541 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTCAGGACTTCTTGGCCCTTGCC[C/T]ATTGCCATACTCTACTGTCAGATTT
Phenotype
MIM: 616637
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101928738 PubMed Links

Gene Details

Gene
LOC101928738
Gene Name
uncharacterized LOC101928738
There are no transcripts associated with this gene.

Gene
LOC101928766
Gene Name
uncharacterized LOC101928766
There are no transcripts associated with this gene.

Gene
TBC1D16
Gene Name
TBC1 domain family member 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271844.1 10673 UTR 3 NP_001258773.1
NM_001271845.1 10673 UTR 3 NP_001258774.1
NM_001271846.1 10673 Intron NP_001258775.1
NM_019020.3 10673 UTR 3 NP_061893.2
XM_005257049.2 10673 UTR 3 XP_005257106.1
XM_005257050.3 10673 UTR 3 XP_005257107.1
XM_005257052.3 10673 Intron XP_005257109.1
XM_006721694.3 10673 UTR 3 XP_006721757.1
XM_006721695.3 10673 Intron XP_006721758.1
XM_011524322.2 10673 UTR 3 XP_011522624.1
XM_011524323.2 10673 UTR 3 XP_011522625.2
XM_011524324.2 10673 UTR 3 XP_011522626.1
XM_017024185.1 10673 Intron XP_016879674.1
XM_017024186.1 10673 UTR 3 XP_016879675.1
XM_017024187.1 10673 UTR 3 XP_016879676.1
XM_017024188.1 10673 UTR 3 XP_016879677.1

View Full Product Details