Product Details
- SNP ID
-
rs2056943
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:15524332 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAGCATGTCAAATCTTTGAGAAGAA[C/T]TCATGACGGAACCACACCACTGTTG
- Phenotype
-
MIM: 607145
MIM: 601594
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DTNBP1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117610176] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DTNBP1
- Gene Name
- dystrobrevin binding protein 1
- Gene
- JARID2
- Gene Name
- jumonji and AT-rich interaction domain containing 2
There are no transcripts associated with this gene.
View Full Product Details