Product Details

SNP ID
rs1815540
Assay Type
Functionally tested
NCBI dbSNP Submissions
38
Location
Chr.1:101873361 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATATATCACAATTTATTTTGTATGG[G/T]GGAAATTGTCTTCAATATATGAAAT
Phenotype
MIM: 607567
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DNAJA1P5 PubMed Links

Gene Details

Gene
DNAJA1P5
Gene Name
DnaJ heat shock protein family (Hsp40) member A1 pseudogene 5
There are no transcripts associated with this gene.

Gene
OLFM3
Gene Name
olfactomedin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288821.1 Intron NP_001275750.1
NM_001288823.1 Intron NP_001275752.1
NM_058170.3 Intron NP_477518.2
XM_017000240.1 Intron XP_016855729.1

View Full Product Details