Product Details

SNP ID
rs677
Assay Type
Validated
NCBI dbSNP Submissions
31
Location
Chr.1:173907423 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TGTTCATGCATCTCCTTTCTGTACC[C/G]TAAGAGAGTGGGGAAGGTGTACTCA
Phenotype
MIM: 107300
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SERPINC1 PubMed Links

Gene Details

Gene
SERPINC1
Gene Name
serpin family C member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000488.3 Intron NP_000479.1
XM_005245198.3 Intron XP_005245255.1

View Full Product Details