Product Details

SNP ID
rs11666
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:87681659 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACTGAGGAGAAATGCATGTGGCT[A/G]TGCATCAGAAGTACAACACGGATCT
Phenotype
MIM: 615165 MIM: 604972
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AKIRIN2 PubMed Links

Gene Details

Gene
AKIRIN2
Gene Name
akirin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018064.3 923 Nonsense Mutation CAG,TAG Q114* NP_060534.1
Gene
ORC3
Gene Name
origin recognition complex subunit 3
There are no transcripts associated with this gene.

View Full Product Details