Product Details

SNP ID
rs1869870
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:46985992 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACCCTGGCTCCCTGAGGGACTCCC[A/G]TACCCCGACACATTGCAGTCTCAAG
Phenotype
MIM: 614169
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC12 PubMed Links

Gene Details

Gene
CCDC12
Gene Name
coiled-coil domain containing 12
There are no transcripts associated with this gene.

Gene
NBEAL2
Gene Name
neurobeachin like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015175.2 Intron NP_055990.1
XM_006713072.3 Intron XP_006713135.1
XM_011533533.2 Intron XP_011531835.1
XM_017006010.1 Intron XP_016861499.1
XM_017006011.1 Intron XP_016861500.1
XM_017006012.1 Intron XP_016861501.1
XM_017006013.1 Intron XP_016861502.1
XM_017006014.1 Intron XP_016861503.1
XM_017006015.1 Intron XP_016861504.1
XM_017006016.1 Intron XP_016861505.1
XM_017006017.1 Intron XP_016861506.1

View Full Product Details