Product Details
- SNP ID
-
rs1979226
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:42993332 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCAGCCTAGCCCTGGCGGCGATTT[C/T]TAGGTCGTAGGAAGTCGATGTTTCG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FLJ32255
PubMed Links
Gene Details
- Gene
- FLJ32255
- Gene Name
- uncharacterized LOC643977
There are no transcripts associated with this gene.
View Full Product Details