Product Details
- SNP ID
-
rs1043935
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:98471295 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGACTGGGTCAATGTTGTTTCCAA[A/C]TTGGGTTTAATTCCATTCAAAGAGA
- Phenotype
-
MIM: 613136
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TSPAN5
PubMed Links
Gene Details
- Gene
- TSPAN5
- Gene Name
- tetraspanin 5
View Full Product Details