Product Details

SNP ID
rs1861725
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:103141999 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTGTGGAGCTTTTTAAACTACACC[A/C]GAGGCTACTGAATCAAAATGTCAGG
Phenotype
MIM: 612334
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NAPEPLD PubMed Links

Gene Details

Gene
NAPEPLD
Gene Name
N-acyl phosphatidylethanolamine phospholipase D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122838.1 Intron NP_001116310.1
NM_198990.4 Intron NP_945341.3
XM_005250214.4 Intron XP_005250271.2
XM_005250215.2 Intron XP_005250272.1
XM_005250218.2 Intron XP_005250275.1
XM_011515933.2 Intron XP_011514235.1
XM_011515934.2 Intron XP_011514236.1
XM_017011854.1 Intron XP_016867343.1
XM_017011855.1 Intron XP_016867344.1
XM_017011856.1 Intron XP_016867345.1
XM_017011857.1 Intron XP_016867346.1
Gene
RPL19P12
Gene Name
ribosomal protein L19 pseudogene 12
There are no transcripts associated with this gene.

View Full Product Details