Product Details

SNP ID
rs12673
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:234391636 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCTGGTGTACTCCCAGATCAGCA[A/G]GGCTCCACTCACATGGACATTCAGG
Phenotype
MIM: 614772 MIM: 605052
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COA6 PubMed Links

Gene Details

Gene
COA6
Gene Name
cytochrome c oxidase assembly factor 6
There are no transcripts associated with this gene.

Gene
TARBP1
Gene Name
TAR (HIV-1) RNA binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005646.3 3420 Silent Mutation CTG,TTG L1603L NP_005637.3
XM_005273234.3 3420 Silent Mutation CTG,TTG L1602L XP_005273291.1
XM_011544263.2 3420 Silent Mutation CTG,TTG L1579L XP_011542565.1
XM_017002194.1 3420 Intron XP_016857683.1
XM_017002195.1 3420 Silent Mutation CTG,TTG L1147L XP_016857684.1
XM_017002196.1 3420 Silent Mutation CTG,TTG L860L XP_016857685.1
XM_017002197.1 3420 Silent Mutation CTG,TTG L806L XP_016857686.1

View Full Product Details