Product Details

SNP ID
rs1317329
Assay Type
Functionally tested
NCBI dbSNP Submissions
48
Location
Chr.1:23640347 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACATAGGTGTTCAATAAATGACAGA[C/T]GCTACCATCACAGATTACCAATGTT
Phenotype
MIM: 607305
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MDS2 PubMed Links

Gene Details

Gene
MDS2
Gene Name
myelodysplastic syndrome 2 translocation associated
There are no transcripts associated with this gene.

View Full Product Details