Product Details

SNP ID
rs14945
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:19975624 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATGCTTTCTGCATGGCCAAGGAA[A/C]TTGGTTAGGGTAGGGAGGGAGGGTA
Phenotype
MIM: 172411
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PLA2G2A PubMed Links

Gene Details

Gene
PLA2G2A
Gene Name
phospholipase A2 group IIA
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000300.3 696 UTR 3 NP_000291.1
NM_001161727.1 696 UTR 3 NP_001155199.1
NM_001161728.1 696 UTR 3 NP_001155200.1
NM_001161729.1 696 UTR 3 NP_001155201.1

View Full Product Details