Product Details

SNP ID
rs15539
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:228488992 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGGTCTGTGGAAATCATGAGAAA[G/T]GACTTGAATGACGCCCGGGACCTGC
Phenotype
MIM: 613754
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RNF187 PubMed Links

Gene Details

Gene
RNF187
Gene Name
ring finger protein 187
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010858.2 545 Missense Mutation AAG,AAT K141N NP_001010858.2

View Full Product Details