Product Details

SNP ID
rs1138267
Assay Type
Functionally tested
NCBI dbSNP Submissions
48
Location
Chr.1:18872082 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTACAGGTAGGGCCTGAGGATCA[A/G]AGGGAAGGCTGACATAGGACGAAGG
Phenotype
MIM: 606811
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALDH4A1 PubMed Links

Gene Details

Gene
ALDH4A1
Gene Name
aldehyde dehydrogenase 4 family member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161504.1 2731 UTR 3 NP_001154976.1
NM_001319218.1 2731 UTR 3 NP_001306147.1
NM_003748.3 2731 UTR 3 NP_003739.2
NM_170726.2 2731 Intron NP_733844.1

View Full Product Details