Product Details

SNP ID
rs1971256
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:151494876 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACAAGAAGATCTGGCTGGCTCAG[C/T]CTGGGTTAAGAATCCCCCTTTGGCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC170 PubMed Links

Gene Details

Gene
CCDC170
Gene Name
coiled-coil domain containing 170
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025059.3 Intron NP_079335.2
XM_011536147.2 Intron XP_011534449.1
XM_011536148.2 Intron XP_011534450.1

View Full Product Details